accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2

CAS RN 61424-26-8

IUPAC Name: 3-benzylaniline
Synonyms: 3-benzylaniline
Molecular Weight (g/mol): 183.25
Molecular Formula: C13H13N
InChi Key: GZYMMTQDXHJALZ-UHFFFAOYSA-N
SMILES: NC1=CC=CC(CC2=CC=CC=C2)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results