accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOOO

CAS RN 61424-76-8

IUPAC Name: 2-amino-4-oxo-4H-chromene-3-carbaldehyde
Synonyms: 2-amino-4-oxochromene-3-carbaldehyde
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: TVGIYZVZBKAJRR-UHFFFAOYSA-N
SMILES: NC1=C(C=O)C(=O)C2=CC=CC=C2O1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results