accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)OHNHOO

CAS RN 61425-27-2

IUPAC Name: benzyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
Synonyms: benzyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
Molecular Weight (g/mol): 209.25
Molecular Formula: C11H15NO3
InChi Key: AFPHMHSLDRPUSM-SECBINFHSA-N
SMILES: C[C@H](CO)NC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)