accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61445-50-9

IUPAC Name: 4-(2,4-dihydroxybenzoyl)benzene-1,2-diol
Synonyms: 4-(2,4-dihydroxybenzoyl)benzene-1,2-diol
Molecular Weight (g/mol): 246.22
Molecular Formula: C13H10O5
InChi Key: UQQYIAVMUUJWGX-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C(C=C1)C(=O)C1=CC(O)=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results