accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHPOHOPOHOHO

CAS RN 6145-31-9

IUPAC Name: (2-phosphonoethyl)phosphonic acid
Synonyms: 2-phosphonoethylphosphonic acid
Molecular Weight (g/mol): 190.03
Molecular Formula: C2H8O6P2
InChi Key: XYJLPCAKKYOLGU-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCP(O)(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)