accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6147-66-6

IUPAC Name: bis(prop-2-en-1-yl)amine hydrochloride
Synonyms: diallylamine hydrochloride
Molecular Weight (g/mol): 133.62
Molecular Formula: C6H12ClN
InChi Key: PZNOBXVHZYGUEX-UHFFFAOYSA-N
SMILES: Cl.C=CCNCC=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results