accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61471-45-2

IUPAC Name: 3-(1H-pyrrol-1-yl)benzoate
Synonyms: 3-(pyrrol-1-yl)benzoate
Molecular Weight (g/mol): 186.19
Molecular Formula: C11H8NO2
InChi Key: PODFNQCZFHLJPH-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(=C1)N1C=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results