accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61477-40-5

IUPAC Name: 3-aminobutan-1-ol
Synonyms: 3-aminobutan-1-ol
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: AGMZSYQMSHMXLT-UHFFFAOYNA-N
SMILES: CC(N)CCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results