accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61495-04-3

IUPAC Name: 2,2-dimethyl-N-(2-methylphenyl)propanamide
Synonyms: 2,2-dimethyl-N-(2-methylphenyl)propanamide
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: CSGRQLUGMVFNON-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NC(=O)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)