accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 615-13-4

IUPAC Name: 2,3-dihydro-1H-inden-2-one
Synonyms: 2-indanone β-hydrindone
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: UMJJFEIKYGFCAT-UHFFFAOYSA-N
SMILES: O=C1CC2=CC=CC=C2C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)