accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 615-16-7

IUPAC Name: 2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 2-benzimidazolinone|O-phenylene urea
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: SILNNFMWIMZVEQ-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2N1
Percent Purity 
  • (1)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (6)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)