accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NHNSN

CAS RN 615-21-4

IUPAC Name: 2-hydrazinyl-1,3-benzothiazole
Synonyms: 2-hydrazinobenzothiazole
Molecular Weight (g/mol): 165.21
Molecular Formula: C7H7N3S
InChi Key: JYSUYJCLUODSLN-UHFFFAOYSA-N
SMILES: NNC1=NC2=CC=CC=C2S1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)