accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 615-50-9

IUPAC Name: 2-methylbenzene-1,4-diamine; sulfuric acid
Synonyms: toluene-2,5-diamine sulfate
Molecular Weight (g/mol): 220.24
Molecular Formula: C7H12N2O4S
InChi Key: KZTWOUOZKZQDMN-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.CC1=CC(N)=CC=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results