accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrBr

CAS RN 615-59-8

IUPAC Name: 1,4-dibromo-2-methylbenzene
Synonyms: benzene, 1,4-dibromo-2-methyl-
Molecular Weight (g/mol): 249.93
Molecular Formula: C7H6Br2
InChi Key: QKEZTJYRBHOKHH-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (5)
  • (1)
Quantity 
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)