accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHCl

CAS RN 615-62-3

IUPAC Name: 3-chloro-4-methylphenol
Synonyms: phenol, 3-chloro-4-methyl-
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: VQZRLBWPEHFGCD-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C=C1Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)