accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3O

CAS RN 615-79-2

IUPAC Name: ethyl 2,4-dioxopentanoate
Synonyms: ethyl 2,4-dioxopentanoate
Molecular Weight (g/mol): 158.15
Molecular Formula: C7H10O4
InChi Key: OYQVQWIASIXXRT-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)CC(C)=O
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)