accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NH2ONHOHO

CAS RN 615-82-7

IUPAC Name: 2-(2-amino-4-methylpentanamido)acetic acid
Synonyms: (2-amino-4-methylpentanamido)acetic acid
Molecular Weight (g/mol): 188.23
Molecular Formula: C8H16N2O3
InChi Key: LESXFEZIFXFIQR-UHFFFAOYNA-N
SMILES: CC(C)CC(N)C(=O)NCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results