accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBrOOCH3

CAS RN 615-83-8

IUPAC Name: ethyl 2-bromopentanoate
Synonyms: ethyl 2-bromopentanoate
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: ORSIRXYHFPHWTN-UHFFFAOYNA-N
SMILES: CCCC(Br)C(=O)OCC
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)