accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OHOOHOOOHOHOH

CAS RN 6151-25-3

IUPAC Name: 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate
Synonyms: quercetin dihydrate
Molecular Weight (g/mol): 338.27
Molecular Formula: C15H14O9
InChi Key: GMGIWEZSKCNYSW-UHFFFAOYSA-N
SMILES: O.O.OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1
Percent Purity 
  • (2)
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (5)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)