accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNCH3Br

CAS RN 61545-99-1

IUPAC Name: 3-methyl-1-octyl-1H-imidazol-3-ium bromide
Synonyms: 1-octyl-3-methylimidazolium bromide
Molecular Weight (g/mol): 275.23
Molecular Formula: C12H23BrN2
InChi Key: URVSXZLUUCVGQM-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results