accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 615534-73-1

IUPAC Name: 1-[(3-iodophenyl)methyl]-1H-1,2,4-triazole
Synonyms: 1-[(3-iodophenyl)methyl]-1,2,4-triazole
Molecular Weight (g/mol): 285.09
Molecular Formula: C9H8IN3
InChi Key: RFHFJBONYNRFAA-UHFFFAOYSA-N
SMILES: IC1=CC=CC(CN2C=NC=N2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results