CAS RN 6156-78-1
Manganese(II) acetate tetrahydrate, 99+%, for analysis
CAS: 6156-78-1 Molecular Formula: C4H14MnO8 Molecular Weight (g/mol): 245.09 MDL Number: MFCD00062552 InChI Key: CESXSDZNZGSWSP-UHFFFAOYSA-L Synonym: manganese ii acetate tetrahydrate,manganese acetate tetrahydrate,manganous acetate tetrahydrate,unii-9to51d176n,manganese diacetate, tetrahydrate,manganese 2+ diacetate tetrahydrate,acetic acid, manganese 2+ salt, tetrahydrate,manganese ii acetatetetrahydrate,acmc-20akkp PubChem CID: 93021 SMILES: O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Manganese acetate tetrahydrate, 99.999%, (trace metal basis)
CAS: 6156-78-1 Molecular Formula: C4H14MnO8 Molecular Weight (g/mol): 245.09 MDL Number: MFCD00062552 InChI Key: CESXSDZNZGSWSP-UHFFFAOYSA-L Synonym: manganese ii acetate tetrahydrate,manganese acetate tetrahydrate,manganous acetate tetrahydrate,unii-9to51d176n,manganese diacetate, tetrahydrate,manganese 2+ diacetate tetrahydrate,acetic acid, manganese 2+ salt, tetrahydrate,manganese ii acetatetetrahydrate,acmc-20akkp PubChem CID: 93021 IUPAC Name: manganese(2+);diacetate;tetrahydrate SMILES: O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)
CAS: 6156-78-1 Molecular Formula: C4H14MnO8 Molecular Weight (g/mol): 245.09 MDL Number: MFCD00062552 InChI Key: CESXSDZNZGSWSP-UHFFFAOYSA-L Synonym: manganese ii acetate tetrahydrate,manganese acetate tetrahydrate,manganous acetate tetrahydrate,unii-9to51d176n,manganese diacetate, tetrahydrate,manganese 2+ diacetate tetrahydrate,acetic acid, manganese 2+ salt, tetrahydrate,manganese ii acetatetetrahydrate,acmc-20akkp PubChem CID: 93021 IUPAC Name: manganese(2+);diacetate;tetrahydrate SMILES: O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Manganese(II) acetate tetrahydrate, Mn 22% (typical)
CAS: 6156-78-1 Molecular Formula: C4H14MnO8 Molecular Weight (g/mol): 245.09 MDL Number: MFCD00062552 InChI Key: CESXSDZNZGSWSP-UHFFFAOYSA-L Synonym: manganese ii acetate tetrahydrate,manganese acetate tetrahydrate,manganous acetate tetrahydrate,unii-9to51d176n,manganese diacetate, tetrahydrate,manganese 2+ diacetate tetrahydrate,acetic acid, manganese 2+ salt, tetrahydrate,manganese ii acetatetetrahydrate,acmc-20akkp PubChem CID: 93021 IUPAC Name: manganese(2+);diacetate;tetrahydrate SMILES: O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Manganese AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Mn), LabChem™
CAS: 6156-78-1 Molecular Formula: C4H14MnO8 Molecular Weight (g/mol): 245.09 MDL Number: MFCD00062552 InChI Key: CESXSDZNZGSWSP-UHFFFAOYSA-L PubChem CID: 93021 IUPAC Name: manganese(2+) diacetate tetrahydrate SMILES: O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Manganese ICP Standard, 10,000 ppm Mn in 5% HNO3, Ricca Chemical
CAS: 6156-78-1 Molecular Formula: C4H14MnO8 Molecular Weight (g/mol): 245.09 MDL Number: MFCD00062552 InChI Key: CESXSDZNZGSWSP-UHFFFAOYSA-L PubChem CID: 93021 IUPAC Name: manganese(2+) diacetate tetrahydrate SMILES: O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Manganese ICP Standard, 1000 ppm Mn in 3% HNO3, Ricca Chemical
CAS: 6156-78-1 Molecular Formula: C4H14MnO8 Molecular Weight (g/mol): 245.09 MDL Number: MFCD00062552 InChI Key: CESXSDZNZGSWSP-UHFFFAOYSA-L PubChem CID: 93021 IUPAC Name: manganese(2+) diacetate tetrahydrate SMILES: O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Manganese ICP-MS Standard, 1000 ppm Mn in 3% HNO3, Ricca Chemical
CAS: 6156-78-1 Molecular Formula: C4H14MnO8 Molecular Weight (g/mol): 245.09 MDL Number: MFCD00062552 InChI Key: CESXSDZNZGSWSP-UHFFFAOYSA-L PubChem CID: 93021 IUPAC Name: manganese(2+) diacetate tetrahydrate SMILES: O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Sigma Aldrich Manganese(II) acetate tetrahydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Manganese(II) acetate tetrahydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.