accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOH

CAS RN 61563-43-7

IUPAC Name: isoquinoline-8-carboxylic acid
Synonyms: isoquinoline-8-carboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: VMNZQPRIUSJCOH-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=NC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results