accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3H3C

CAS RN 61566-34-5

IUPAC Name: methyl 2-[4-(2-methylpropyl)phenyl]propanoate
Synonyms: methyl 2-[4-(2-methylpropyl)phenyl]propanoate
Molecular Weight (g/mol): 220.31
Molecular Formula: C14H20O2
InChi Key: YNZYUHPFNYBBFF-UHFFFAOYNA-N
SMILES: COC(=O)C(C)C1=CC=C(CC(C)C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results