accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 6158-45-8

IUPAC Name: 1-(propan-2-yl)naphthalene
Synonyms: 1-isopropylnaphthalene
Molecular Weight (g/mol): 170.26
Molecular Formula: C13H14
InChi Key: PMPBFICDXLLSRM-UHFFFAOYSA-N
SMILES: CC(C)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results