accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6159-05-3

IUPAC Name: 1,1'-diheptyl-[4,4'-bipyridine]-1,1'-diium dibromide
Synonyms: 1,1'-diheptyl-[4,4'-bipyridine]-1,1'-diium dibromide
Molecular Weight (g/mol): 514.39
Molecular Formula: C24H38Br2N2
InChi Key: VRXAJMCFEOESJO-UHFFFAOYSA-L
SMILES: [Br-].[Br-].CCCCCCC[N+]1=CC=C(C=C1)C1=CC=[N+](CCCCCCC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results