accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OCH3OHUOHH3COOOO

CAS RN 6159-44-0

IUPAC Name: uranyl acetate
Synonyms: uranyl acetate
Molecular Weight (g/mol): 426.16
Molecular Formula: C4H12O8U
InChi Key: COQLPRJCUIATTQ-UHFFFAOYSA-N
SMILES: O.O.O=[U]=O.CC(O)=O.CC(O)=O
Grade 
  • (6)
  • (3)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)