accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3C(E)

CAS RN 616-12-6

IUPAC Name: (2E)-3-methylpent-2-ene
Synonyms: trans-3-methyl-2-pentene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: BEQGRRJLJLVQAQ-GQCTYLIASA-N
SMILES: CC\C(C)=C\C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)