accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClCl

CAS RN 616-21-7

IUPAC Name: 1,2-dichlorobutane
Synonyms: 1,2-dichlorobutane
Molecular Weight (g/mol): 127.01
Molecular Formula: C4H8Cl2
InChi Key: PQBOTZNYFQWRHU-UHFFFAOYNA-N
SMILES: CCC(Cl)CCl
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)