accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HOH2N

CAS RN 616-29-5

IUPAC Name: 1,3-diaminopropan-2-ol
Synonyms: 1,3-diaminopropan-2-ol
Molecular Weight (g/mol): 90.13
Molecular Formula: C3H10N2O
InChi Key: UYBWIEGTWASWSR-UHFFFAOYSA-N
SMILES: NCC(O)CN
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (4)
  • (5)
  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)