accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 616-39-7

IUPAC Name: diethyl(methyl)azanium
Synonyms: diethyl(methyl)azanium
Molecular Weight (g/mol): 88.17
Molecular Formula: C5H14N
InChi Key: GNVRJGIVDSQCOP-UHFFFAOYSA-O
SMILES: CC[NH+](C)CC
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)