accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 616-44-4

IUPAC Name: 3-methylthiophene
Synonyms: 3-methylthiophene
Molecular Weight (g/mol): 98.16
Molecular Formula: C5H6S
InChi Key: QENGPZGAWFQWCZ-UHFFFAOYSA-N
SMILES: CC1=CSC=C1
Molecular Weight (g/mol) 
  • (6)
  • (4)
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (2)
  • (5)
  • (6)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (7)
  • (1)