accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 616-79-5

IUPAC Name: 2-amino-5-nitrobenzoic acid
Synonyms: 2-amino-5-nitrobenzoic acid
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H6N2O4
InChi Key: RUCHWTKMOWXHLU-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1C(O)=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results