accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3COCH3H3C

CAS RN 61676-61-7

IUPAC Name: 4,4,6-trimethyl-2-(propan-2-yloxy)-1,3,2-dioxaborinane
Synonyms: 2-isopropoxy-4,4,6-trimethyl-1,3,2-dioxaborinane
Molecular Weight (g/mol): 186.06
Molecular Formula: C9H19BO3
InChi Key: AHCZPHINTQYKPU-UHFFFAOYNA-N
SMILES: CC(C)OB1OC(C)CC(C)(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results