CAS RN 61676-62-8
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 94%
CAS: 61676-62-8 Molecular Formula: C9H19BO3 Molecular Weight (g/mol): 186.058 MDL Number: MFCD00192241 InChI Key: MRWWWZLJWNIEEJ-UHFFFAOYSA-N Synonym: 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,isopropoxyboronic acid pinacol ester,4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane,pinbop,isopropoxyboronic acid, pinacol ester,isopropyl pinacol borate,2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane,2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane,2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane PubChem CID: 5017741 IUPAC Name: 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)OC(C)C
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
CAS: 61676-62-8 Molecular Formula: C9H19BO3 Molecular Weight (g/mol): 186.058 MDL Number: MFCD00192241 InChI Key: MRWWWZLJWNIEEJ-UHFFFAOYSA-N Synonym: 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,isopropoxyboronic acid pinacol ester,4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane,pinbop,isopropoxyboronic acid, pinacol ester,isopropyl pinacol borate,2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane,2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane,2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane PubChem CID: 5017741 IUPAC Name: 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)OC(C)C
Sigma Aldrich 2-Isopropoxy-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.