accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6169-06-8

IUPAC Name: octan-2-ol
Synonyms: 2-octanol sec-caprylic alcohol s-octyl alcohol 2-octanol (natural) secondary caprylic alcohol fema number 2801 caprylic alcohol, secondary
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: SJWFXCIHNDVPSH-UHFFFAOYNA-N
SMILES: CCCCCCC(C)O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)