accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONHNHO

CAS RN 61698-32-6

IUPAC Name: [5-(hydroxymethyl)-2-phenyl-1H-imidazol-4-yl]methanol
Synonyms: [5-(hydroxymethyl)-2-phenyl-1H-imidazol-4-yl]methanol
Molecular Weight (g/mol): 204.23
Molecular Formula: C11H12N2O2
InChi Key: UUQQGGWZVKUCBD-UHFFFAOYSA-N
SMILES: OCC1=C(CO)N=C(N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results