CAS RN 617-29-8
CAS RN 617-29-8
IUPAC Name:
2-methylhexan-3-ol
Synonyms:
2-methyl-3-hexanol
Molecular Weight (g/mol):
116.2
Molecular Formula:
C7H16O
InChi Key:
RGRUUTLDBCWYBL-UHFFFAOYNA-N
SMILES:
CCCC(O)C(C)C
1
–
1
of
1
results
1
2-Methyl-3-hexanol, TCI America™
CAS: 617-29-8 Molecular Formula: C7H16O Molecular Weight (g/mol): 116.204 MDL Number: MFCD00004577 InChI Key: RGRUUTLDBCWYBL-UHFFFAOYSA-N PubChem CID: 12040 IUPAC Name: 2-methylhexan-3-ol SMILES: CCCC(C(C)C)O