accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3CH3

CAS RN 617-50-5

IUPAC Name: propan-2-yl 2-methylpropanoate
Synonyms: isopropyl isobutyrate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: WVRPFQGZHKZCEB-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results