accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH2N

CAS RN 617-79-8

IUPAC Name: 3-(aminomethyl)pentane
Synonyms: 2-ethylbutylamine
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: MGWAGIQQTULHGU-UHFFFAOYSA-N
SMILES: CCC(CC)CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results