accessibility menu, dialog, popup

UserName

Viewing profile as user:
NO

CAS RN 617-90-3

IUPAC Name: furan-2-carbonitrile
Synonyms: 2-furonitrile
Molecular Weight (g/mol): 93.09
Molecular Formula: C5H3NO
InChi Key: YXDXXGXWFJCXEB-UHFFFAOYSA-N
SMILES: N#CC1=CC=CO1
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (6)