accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNHN

CAS RN 61700-61-6

IUPAC Name: 1H-indazole-5-carboxylic acid
Synonyms: 1H-indazole-5-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: MAVGBUDLHOOROM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2NN=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results