accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61702-43-0

IUPAC Name: sodium 2-amino-4-nitrobenzen-1-olate
Synonyms: sodium 2-amino-4-nitrobenzenolate
Molecular Weight (g/mol): 176.11
Molecular Formula: C6H5N2NaO3
InChi Key: QROXVHUAKDVYQE-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC(=CC=C1[O-])[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results