accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61715-72-8

IUPAC Name: 1-ethyl-1-(4-methylphenyl)hydrazine
Synonyms: 1-ethyl-1-(4-methylphenyl)hydrazine
Molecular Weight (g/mol): 150.23
Molecular Formula: C9H14N2
InChi Key: IFARRRGGHHADCF-UHFFFAOYSA-N
SMILES: CCN(N)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results