accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61751-44-8

IUPAC Name: 9-methyl-2H,3H,4H-pyrido[1,2-a]pyrimidin-2-one
Synonyms: 9-methyl-3H,4H-pyrido[1,2-a]pyrimidin-2-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C9H10N2O
InChi Key: HPVULYKIYNGGJG-UHFFFAOYSA-N
SMILES: CC1=CC=CN2CCC(=O)N=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results