accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 618-21-3

IUPAC Name: 4-phosphonobenzoic acid
Synonyms: 4-phosphonobenzoic acid
Molecular Weight (g/mol): 202.1
Molecular Formula: C7H7O5P
InChi Key: IEQICHVXWFGDAN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)P(O)(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results