accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)O(S)OO(R)OH(S)OH(S)OH

CAS RN 618-65-5

IUPAC Name: 2-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}benzaldehyde
Synonyms: helicin
Molecular Weight (g/mol): 284.26
Molecular Formula: C13H16O7
InChi Key: BGOFCVIGEYGEOF-UJPOAAIJSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC=CC=C2C=O)[C@H](O)[C@@H](O)[C@@H]1O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)