accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOI

CAS RN 618-91-7

IUPAC Name: methyl 3-iodobenzoate
Synonyms: methyl 3-iodobenzoate
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: NPXOIGSBRLCOSD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results