accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 6180-61-6

IUPAC Name: 3-phenoxypropan-1-ol
Synonyms: 3-phenoxypropan-1-ol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: AWVDYRFLCAZENH-UHFFFAOYSA-N
SMILES: OCCCOC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)