accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(S)(S)OOH3CCH3

CAS RN 61826-55-9

IUPAC Name: 2-[(1S,3S)-3-acetyl-2,2-dimethylcyclobutyl]acetate
Synonyms: 2-[(1S,3S)-3-acetyl-2,2-dimethylcyclobutyl]acetate
Molecular Weight (g/mol): 183.23
Molecular Formula: C10H15O3
InChi Key: SIZDUQQDBXJXLQ-JGVFFNPUSA-M
SMILES: CC(=O)[C@H]1C[C@@H](CC([O-])=O)C1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results